Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111169.1 | NP_001104639.1 | 8 - 9 | 790 | 91 | |
XM_005160743.4 | XP_005160800.1 | 7 - 8 | 857 | 91 | ||
XM_005160744.4 | XP_005160801.1 | 7 - 8 | 857 | 91 | ||
XM_009294764.3 | XP_009293039.1 | 8 - 9 | 899 | 91 | ||
GenBank mRNA | EU245032.1 | - | 8 - 9 | 790 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Danio rerio Probe spans exons |