Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: olfm3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111172.1 | NP_001104642.1 | 2 - 3 | 509 | 90 | |
GenBank mRNA | BC153993.1 | - | 2 - 3 | 509 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |