Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: htra1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111182.1 | NP_001104652.1 | 2 - 3 | 684 | 63 | |
GenBank mRNA | BC155591.1 | - | 2 - 3 | 684 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |