Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: and2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111190.1 | NP_001104660.1 | 2 - 3 | 141 | 79 | |
GenBank mRNA | BC155829.1 | - | 2 - 3 | 141 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |