Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: abcg4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111212.1 | NP_001104682.1 | 7 - 8 | 855 | 90 | |
GenBank mRNA | BC153997.1 | - | 7 - 8 | 855 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |