Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spint1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111223.1 | NP_001104693.1 | 8 - 9 | 1463 | 70 | |
GenBank mRNA | BC155551.1 | - | 8 - 9 | 1463 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |