Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spred3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113215.2 | NP_001106686.2 | 1 - 2 | 46 | 74 | |
XM_005159247.4 | XP_005159304.1 | 1 - 2 | 344 | 74 | ||
GenBank mRNA | BC155342.1 | - | 1 - 2 | 46 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |