Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spred3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113215.2 | NP_001106686.2 | 3 - 4 | 374 | 95 | |
XM_005159247.4 | XP_005159304.1 | 3 - 4 | 672 | 95 | ||
XM_021467687.1 | XP_021323362.1 | 3 - 4 | 463 | 95 | ||
XM_021467688.1 | XP_021323363.1 | 2 - 3 | 290 | 95 | ||
XM_021467689.1 | XP_021323364.1 | 3 - 4 | 463 | 95 | ||
GenBank mRNA | BC155342.1 | - | 3 - 4 | 374 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |