Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp27c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113337.3 | NP_001106808.2 | 6 - 7 | 1355 | 65 | |
GenBank mRNA | BC154632.1 | - | 6 - 7 | 1264 | 65 | |
BC171576.1 | - | 6 - 7 | 1040 | 65 | ||
BC171578.1 | - | 6 - 7 | 1040 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |