Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prlrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113500.1 | NP_001106972.1 | 3 - 4 | 382 | 72 | |
XM_005165610.3 | XP_005165667.1 | 4 - 5 | 588 | 72 | ||
GenBank mRNA | EU517720.1 | - | 4 - 5 | 560 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |