Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nos2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113501.1 | NP_001106973.1 | 12 - 13 | 1602 | 93 | |
XM_005157694.4 | XP_005157751.1 | 4 - 5 | 365 | 93 | ||
XM_009291655.3 | XP_009289930.1 | 11 - 12 | 2659 | 93 | ||
XM_017350981.2 | XP_017206470.1 | 11 - 12 | 2657 | 93 | ||
GenBank mRNA | AM749802.1 | - | 12 - 13 | 1602 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Danio rerio Probe spans exons |