Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nos2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113501.1 | NP_001106973.1 | 22 - 23 | 2925 | 60 | |
XM_005157694.4 | XP_005157751.1 | 14 - 15 | 1688 | 60 | ||
XM_009291655.3 | XP_009289930.1 | 21 - 22 | 3960 | 60 | ||
XM_017350981.2 | XP_017206470.1 | 21 - 22 | 3980 | 60 | ||
GenBank mRNA | AM749802.1 | - | 22 - 23 | 2925 | 60 | |
EU332350.1 | - | 22 - 23 | 2776 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Danio rerio Probe spans exons |