Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ghrhr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113503.1 | NP_001106975.1 | 4 - 5 | 367 | 65 | |
GenBank mRNA | EU056169.1 | - | 4 - 5 | 367 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Recommended Assay Danio rerio Probe spans exons |