Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ghrhr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113503.1 | NP_001106975.1 | 6 - 7 | 598 | 74 | |
GenBank mRNA | EU056169.1 | - | 6 - 7 | 598 | 74 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Danio rerio Probe spans exons |