Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: il23r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113506.1 | NP_001106978.1 | 7 - 8 | 1090 | 106 | |
XM_021476745.1 | XP_021332420.1 | 10 - 11 | 1972 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |