Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cacnb4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113519.2 | NP_001106991.1 | 1 - 2 | 181 | 61 | |
GenBank mRNA | BC171593.1 | - | 1 - 2 | 87 | 61 | |
BC171597.1 | - | 1 - 2 | 87 | 61 | ||
EF601061.1 | - | 1 - 2 | 185 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |