Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: anxa5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113582.1 | NP_001107054.1 | 5 - 6 | 362 | 94 | |
GenBank mRNA | BC154565.1 | - | 5 - 6 | 362 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |