Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: anapc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113585.1 | NP_001107057.1 | 10 - 11 | 1511 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Danio rerio Probe spans exons |