Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: anapc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113585.1 | NP_001107057.1 | 6 - 7 | 821 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Danio rerio Probe spans exons |