Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccl20b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113595.1 | NP_001107067.1 | 3 - 4 | 252 | 83 | |
GenBank mRNA | AB331755.1 | - | 3 - 4 | 252 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Danio rerio Probe spans exons |