Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tfb2m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113617.2 | NP_001107089.2 | 8 - 9 | 1265 | 99 | |
GenBank mRNA | BC154809.1 | - | 8 - 9 | 1224 | 99 | |
BC171547.1 | - | 8 - 9 | 1188 | 99 | ||
EH461916.1 | - | 1 - 2 | 672 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |