Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sgk2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113622.1 | NP_001107094.1 | 11 - 12 | 1022 | 105 | |
XM_009296592.3 | XP_009294867.1 | 11 - 12 | 1082 | 105 | ||
GenBank mRNA | BC154535.1 | - | 11 - 12 | 1022 | 105 | |
BC171486.1 | - | 11 - 12 | 989 | 105 | ||
BC171490.1 | - | 11 - 12 | 989 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |