Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ankrd39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113640.1 | NP_001107112.1 | 3 - 4 | 395 | 81 | |
XM_005167331.3 | XP_005167388.1 | 4 - 5 | 468 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Danio rerio Probe spans exons |