Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: il6r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001330258.2 | NP_001317187.2 | 9 - 10 | 1987 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Danio rerio Probe spans exons |