Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ncalda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114410.1 | NP_001107882.1 | 4 - 5 | 662 | 63 | |
XM_005173483.4 | XP_005173540.1 | 3 - 4 | 723 | 63 | ||
GenBank mRNA | BC155337.1 | - | 4 - 5 | 662 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |