Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: csf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114480.1 | NP_001107952.1 | 4 - 5 | 479 | 78 | |
GenBank mRNA | AM901598.1 | - | 4 - 5 | 576 | 78 | |
BC155620.1 | - | 4 - 5 | 479 | 78 | ||
KM655659.1 | - | 4 - 5 | 381 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |