Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131241.1 | NP_571316.1 | 5 - 6 | 736 | 106 | |
XM_021468750.1 | XP_021324425.1 | 5 - 6 | 783 | 106 | ||
GenBank mRNA | AF047412.1 | - | 3 - 4 | 434 | 106 | |
BC076051.1 | - | 5 - 6 | 736 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |