Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309822.1 | NP_001296751.1 | 3 - 4 | 376 | 78 | |
NM_131266.2 | NP_571341.2 | 3 - 4 | 376 | 78 | ||
GenBank mRNA | AF191559.1 | - | 3 - 4 | 307 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Danio rerio Probe spans exons |