Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309822.1 | NP_001296751.1 | 7 - 8 | 981 | 83 | |
NM_131266.2 | NP_571341.2 | 7 - 8 | 981 | 83 | ||
GenBank mRNA | AF191559.1 | - | 7 - 8 | 912 | 83 | |
BC059803.1 | - | 7 - 8 | 836 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |