Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: emx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131280.2 | NP_571355.2 | 1 - 2 | 593 | 115 | |
GenBank mRNA | BC092713.1 | - | 1 - 2 | 593 | 115 | |
BC165103.1 | - | 1 - 2 | 424 | 115 | ||
D32215.1 | - | 1 - 2 | 427 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |