Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fgf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131291.1 | NP_571366.1 | 1 - 2 | 277 | 81 | |
GenBank mRNA | BC162856.1 | - | 1 - 2 | 319 | 81 | |
BC162895.1 | - | 1 - 2 | 319 | 81 | ||
KM655696.1 | - | 1 - 2 | 277 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |