Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kdrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131472.1 | NP_571547.1 | 21 - 22 | 2892 | 123 | |
XM_009291142.3 | XP_009289417.1 | 21 - 22 | 3286 | 123 | ||
GenBank mRNA | AF180354.1 | - | 18 - 19 | 2508 | 123 | |
AF487829.1 | - | 21 - 22 | 3245 | 123 | ||
AY056466.1 | - | 21 - 22 | 2892 | 123 | ||
BC129158.1 | - | 21 - 22 | 3260 | 123 | ||
CU458916.1 | - | 21 - 22 | 3260 | 123 |
Gene Symbol: LOC103909948 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009298309.3 | XP_009296584.3 | 3 - 4 | 401 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |