Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kdrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131472.1 | NP_571547.1 | 4 - 5 | 488 | 83 | |
GenBank mRNA | AF180354.1 | - | 1 - 2 | 98 | 83 | |
AF487829.1 | - | 4 - 5 | 838 | 83 | ||
AY056466.1 | - | 4 - 5 | 488 | 83 | ||
BC129158.1 | - | 4 - 5 | 853 | 83 | ||
CU458916.1 | - | 4 - 5 | 853 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |