Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnajc3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131630.1 | NP_571705.1 | 12 - 13 | 1484 | 101 | |
GenBank mRNA | AF295376.1 | - | 12 - 13 | 1484 | 101 | |
BC163418.1 | - | 11 - 12 | 1395 | 101 | ||
BC163421.1 | - | 11 - 12 | 1395 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |