Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnajc3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131630.1 | NP_571705.1 | 7 - 8 | 854 | 82 | |
GenBank mRNA | AF295376.1 | - | 7 - 8 | 854 | 82 | |
BC163418.1 | - | 6 - 7 | 765 | 82 | ||
BC163421.1 | - | 6 - 7 | 765 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |