Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cmlc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131692.2 | NP_571767.2 | 1 | 249 | 67 | |
GenBank mRNA | BC083381.1 | - | 1 | 250 | 67 | |
BC164995.1 | - | 1 | 202 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |