Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cldne | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131765.1 | NP_571840.1 | 1 | 552 | 79 | |
GenBank mRNA | AF359425.1 | - | 1 | 552 | 79 | |
BC062846.1 | - | 1 | 551 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Inventoried Both primers and probe map within a single exon |