Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chrm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178301.1 | NP_840086.1 | 1 | 850 | 95 | |
GenBank mRNA | BC081410.1 | - | 2 | 1042 | 95 | |
BC163917.1 | - | 1 | 893 | 95 | ||
BC163924.1 | - | 1 | 893 | 95 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |