Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pnx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178302.2 | NP_840087.1 | 1 - 2 | 180 | 127 | |
GenBank mRNA | AB067731.1 | - | 1 - 2 | 180 | 127 | |
BC162069.1 | - | 1 - 2 | 133 | 127 | ||
BC162454.1 | - | 1 - 2 | 133 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |