Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hdr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194391.1 | NP_919372.1 | 6 - 7 | 682 | 99 | |
XM_005171798.4 | XP_005171855.1 | 6 - 7 | 1546 | 99 | ||
GenBank mRNA | AF302789.1 | - | 6 - 7 | 682 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Danio rerio Probe spans exons |