Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: depdc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198872.1 | NP_942573.1 | 5 - 6 | 920 | 84 | |
XM_005166773.3 | XP_005166830.1 | 5 - 6 | 921 | 84 | ||
GenBank mRNA | BC044382.1 | - | 5 - 6 | 920 | 84 | |
BC171596.1 | - | 5 - 6 | 872 | 84 | ||
BC171624.1 | - | 5 - 6 | 872 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |