Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atad5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082829.2 | NP_001076298.2 | 2 - 3 | 918 | 142 | |
XM_005166177.4 | XP_005166234.1 | 2 - 3 | 983 | 142 | ||
GenBank mRNA | BC057535.1 | - | 2 - 3 | 918 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |