Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199598.1 | NP_955892.1 | 3 - 4 | 311 | 97 | |
GenBank mRNA | BC045362.1 | - | 3 - 4 | 311 | 97 | |
BC165743.1 | - | 3 - 4 | 241 | 97 | ||
BC166040.1 | - | 3 - 4 | 307 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |