Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199601.1 | NP_955895.1 | 13 - 14 | 1232 | 99 | |
XM_021477397.1 | XP_021333072.1 | 13 - 14 | 1267 | 99 | ||
XM_021477398.1 | XP_021333073.1 | 13 - 14 | 1268 | 99 | ||
GenBank mRNA | BC044366.1 | - | 13 - 14 | 1232 | 99 | |
BC066733.1 | - | 13 - 14 | 1195 | 99 | ||
BC165458.1 | - | 11 - 12 | 1000 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |