Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cpne1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276281.1 | NP_001263210.1 | 12 - 13 | 1195 | 91 | |
NM_199699.2 | NP_955993.2 | 12 - 13 | 1192 | 91 | ||
XM_005169257.4 | XP_005169314.1 | 12 - 13 | 1151 | 91 | ||
XM_005169258.4 | XP_005169315.1 | 12 - 13 | 1147 | 91 | ||
XM_005169260.4 | XP_005169317.1 | 12 - 13 | 1184 | 91 | ||
XM_005169262.3 | XP_005169319.1 | 12 - 13 | 1181 | 91 | ||
XM_009306020.2 | XP_009304295.1 | 12 - 13 | 1147 | 91 | ||
XM_017358205.2 | XP_017213694.1 | 12 - 13 | 1086 | 91 | ||
GenBank mRNA | AY423010.1 | - | 12 - 13 | 1127 | 91 | |
BC050240.1 | - | 12 - 13 | 1185 | 91 | ||
BC151889.1 | - | 12 - 13 | 1154 | 91 | ||
BC164658.1 | - | 11 - 12 | 1095 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |