Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: asb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199770.1 | NP_956064.1 | 2 - 3 | 218 | 70 | |
XM_005162226.4 | XP_005162283.1 | 2 - 3 | 488 | 70 | ||
XM_005162227.4 | XP_005162284.1 | 2 - 3 | 276 | 70 | ||
GenBank mRNA | BC053213.1 | - | 2 - 3 | 218 | 70 | |
BC153632.1 | - | 2 - 3 | 245 | 70 | ||
BC165704.1 | - | 1 - 2 | 164 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |