Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zmpste24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199892.1 | NP_956186.1 | 2 - 3 | 462 | 97 | |
GenBank mRNA | BC046884.1 | - | 2 - 3 | 493 | 97 | |
BC165348.1 | - | 2 - 3 | 288 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |