Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aqp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199910.2 | NP_956204.2 | 1 - 2 | 147 | 67 | |
XM_005161011.4 | XP_005161068.1 | 1 - 2 | 206 | 67 | ||
GenBank mRNA | BC056802.1 | - | 1 - 2 | 223 | 67 | |
FJ655385.1 | - | 1 - 2 | 147 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |