Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aqp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199910.2 | NP_956204.2 | 3 - 4 | 402 | 77 | |
GenBank mRNA | BC056802.1 | - | 3 - 4 | 478 | 77 | |
FJ655385.1 | - | 3 - 4 | 402 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |