Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: polr2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167956.2 | NP_001161428.1 | 7 - 8 | 798 | 81 | |
GenBank mRNA | BC046047.1 | - | 7 - 8 | 710 | 81 | |
EH524379.1 | - | 2 - 3 | 319 | 81 | ||
EH555113.1 | - | 7 - 8 | 842 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |